1,2-Diamino-3,4-difluorobenzene 95%

Supplier: Ambeed
Warning

Synonyms: 3,4-Difluoro-1,2-phenylenediamine, 3,4-Difluorobenzene-1,2-diamine, 3,4-Difluoro-o-phenylenediamine

A1373444-100G A1373444-1G A1373444-25G A1373444-5G
76660-026EA 420 USD
76660-026 76660-028 76660-030 76660-032
1,2-Diamino-3,4-difluorobenzene 95%
1,2-Diamino-3,4-difluorobenzene

Precaución: for Research Use Only

Formula: C₆H₆F₂N₂
MDL Number: MFCD00153128
CAS Number: 153505-39-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR