1-Phenoxy-2-phenylpropan-2-ol 97%

Supplier: Ambeed
Warning

A1373372-1G A1373372-250MG
76660-014EA 274 USD
76660-014 76660-016
1-Phenoxy-2-phenylpropan-2-ol 97%
1-Phenoxy-2-phenylpropan-2-ol

Precaución: for Research Use Only

Formula: C₁₅H₁₆O₂
MW: 228.2863 g/mol
MDL Number: MFCD17966864
CAS Number: 155848-04-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR