(1-Methoxy-2-phenoxyethyl)benzene 95%

Supplier: Ambeed
Warning

A1373370-100MG A1373370-1G A1373370-250MG
76660-008EA 57 USD
76660-008 76660-010 76660-012
(1-Methoxy-2-phenoxyethyl)benzene 95%
(1-Methoxy-2-phenoxyethyl)benzene

Precaución: for Research Use Only

Formula: C₁₅H₁₆O₂
CAS Number: 29509-35-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR