2-Hydroxy-5-(1,2,2-triphenylvinyl)benzaldehyde 97%

Supplier: Ambeed
Warning

A1373322-100MG A1373322-250MG
76660-004EA 112.46 USD
76660-004 76660-006
2-Hydroxy-5-(1,2,2-triphenylvinyl)benzaldehyde 97%
2-Hydroxy-5-(1,2,2-triphenylvinyl)benzaldehyde

Precaución: for Research Use Only

Formula: C₂₇H₂₀O₂
MW: 376.4465 g/mol
CAS Number: 1926206-27-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR