Ethyl 2-(2,6-dimethylphenyl)hydrazinecarboxylate 97%

Supplier: Ambeed
Warning

A1373210-1G A1373210-250MG
76659-990EA 185.66 USD
76659-990 76659-992
Ethyl 2-(2,6-dimethylphenyl)hydrazinecarboxylate 97%
Ethyl 2-(2,6-dimethylphenyl)hydrazinecarboxylate

Precaución: for Research Use Only

Formula: C₁₁H₁₆N₂O₂
MW: 208.2569 g/mol
CAS Number: 112341-87-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR