(R)-2-Hydroxy-2-(p-tolyl)acetic acid 98%

Supplier: Ambeed
Danger

A1373167-1G A1373167-250MG A1373167-5G
76659-984EA 12 USD
76659-984 76659-986 76659-988
(R)-2-Hydroxy-2-(p-tolyl)acetic acid 98%
(R)-2-Hydroxy-2-(p-tolyl)acetic acid

Precaución: for Research Use Only

Formula: C₉H₁₀O₃
MDL Number: MFCD00798433
CAS Number: 31284-89-6
UN: 3261
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR