2-(4-Bromophenyl)quinazolin-4(3H)-one 97%

Supplier: Ambeed
Warning

A1373157-1G A1373157-250MG A1373157-5G
76659-978EA 94.43 USD
76659-978 76659-980 76659-982
2-(4-Bromophenyl)quinazolin-4(3H)-one 97%
2-(4-Bromophenyl)quinazolin-4(3H)-one

Precaución: for Research Use Only

Formula: C₁₄H₉BrN₂O
MW: 301.1381 g/mol
MDL Number: MFCD00571434
CAS Number: 83800-88-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR