6-(Dimethylamino)naphthalene-2-sulfonic acid 97%

Supplier: Ambeed
Warning

A1372884-1G A1372884-250MG
76659-966EA 130.5 USD
76659-966 76659-968
6-(Dimethylamino)naphthalene-2-sulfonic acid 97%
6-(Dimethylamino)naphthalene-2-sulfonic acid

Precaución: for Research Use Only

Formula: C₁₂H₁₃NO₃S
MW: 251.3015 g/mol
CAS Number: 34322-51-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR