4-((2,4-Diaminopyrimidin-5-yl)methyl)-2,6-dimethoxyphenol hydrobromide 97%

Supplier: Ambeed
Warning

A1372882-100MG A1372882-250MG
76659-964EA 66 USD
76659-964 77867-685
4-((2,4-Diaminopyrimidin-5-yl)methyl)-2,6-dimethoxyphenol hydrobromide 97%
4-((2,4-Diaminopyrimidin-5-yl)methyl)-2,6-dimethoxyphenol hydrobromide

Precaución: for Research Use Only

Formula: C₁₃H₁₇BrN₄O₃
MDL Number: MFCD33022474
CAS Number: 2682114-46-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR