4-Bromo-N-(4-nitrophenyl)-N-phenylaniline 97%

Supplier: Ambeed
Warning

A1372806-100MG A1372806-1G
76659-954EA 80 USD
76659-954 76659-956
4-Bromo-N-(4-nitrophenyl)-N-phenylaniline 97%
4-Bromo-N-(4-nitrophenyl)-N-phenylaniline

Precaución: for Research Use Only

Formula: C₁₈H₁₃BrN₂O₂
CAS Number: 1028385-91-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR