Benzothiazole-5-carboxylic acid 95%

Supplier: Ambeed
Warning

A1372794-1G A1372794-250MG A1372794-5G A1372794-25G
76659-948EA 38 USD
76659-948 76659-950 76659-952 77621-926
Benzothiazole-5-carboxylic acid 95%
Benzothiazole-5-carboxylic acid

Precaución: for Research Use Only

Formula: C₈H₅NO₂S
MDL Number: MFCD06796380
CAS Number: 68867-17-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR