2-Amino-4-chloro-5-methylbenzenesulphonic acid sodium salt 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
5-Chloro-4-methylaniline-2-sulphonic acid Sodium Salt, Sodium 2-amino-4-chloro-5-methylbenzenesulphonate, Aminochlorotoluenesulphonicacidsodiumsalt, 5-Chloro-p-toluidine-2-sulphonic acid Sodium Salt, Sodium 2-Amino-4-chloro-5-methylbenzenesulphonate
A1372792-100G
A1372792-25G
A1372792-500G
A1372792-1000G
76659-942EA
23
USD
76659-942
76659-944
76659-946
77621-924
2-Amino-4-chloro-5-methylbenzenesulphonic acid sodium salt 97%
2-Amino-4-chloro-5-methylbenzenesulphonic acid sodium salt
Precaución: for Research Use Only
Formula:
C₇H₇ClNNaO₃S |
MDL Number:
MFCD00070551 CAS Number: 6627-59-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...