1,3-Diisopropyl-1H-imidazol-3-ium iodide 97%

Supplier: Ambeed
Warning

A1372788-100MG A1372788-1G A1372788-250MG
76659-936EA 13 USD
76659-936 76659-938 76659-940
1,3-Diisopropyl-1H-imidazol-3-ium iodide 97%
1,3-Diisopropyl-1H-imidazol-3-ium iodide

Precaución: for Research Use Only

Formula: C₉H₁₇IN₂
CAS Number: 398460-83-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR