3,7-Bis(diethylamino)phenothiazin-5-ium iodide 97%

Supplier: Ambeed
Warning

A1372658-1G A1372658-250MG
76659-916EA 221 USD
76659-916 76659-918
3,7-Bis(diethylamino)phenothiazin-5-ium iodide 97%
3,7-Bis(diethylamino)phenothiazin-5-ium iodide

Precaución: for Research Use Only

Formula: C₂₀H₂₆IN₃S
CAS Number: 439119-93-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR