Formylbis(triphenylphosphoranyl)rhodium(IV) chloride 99%

Supplier: Ambeed
Warning

A1372604-100MG A1372604-1G A1372604-250MG
76659-906EA 46 USD
76659-906 76659-908 76659-910
Formylbis(triphenylphosphoranyl)rhodium(IV) chloride 99%
Formylbis(triphenylphosphoranyl)rhodium(IV) chloride

Precaución: for Research Use Only

Formula: C₃₇H₃₃ClOP₂Rh
MDL Number: MFCD00044273
CAS Number: 13938-94-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR