(S)-2-Amino-2-(4-methoxyphenyl)acetic acid 97%

Supplier: Ambeed
Warning

A1372530-100MG A1372530-250MG
76659-902EA 41.38 USD
76659-902 76659-904
(S)-2-Amino-2-(4-methoxyphenyl)acetic acid 97%
(S)-2-Amino-2-(4-methoxyphenyl)acetic acid

Precaución: for Research Use Only

Formula: C₉H₁₁NO₃
MW: 181.1885 g/mol
MDL Number: MFCD07371762
CAS Number: 24593-48-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR