5-(Aminomethyl)isoindoline-1,3-dione hydrochloride 97%

Supplier: Ambeed
Warning

A1372395-100MG A1372395-1G A1372395-250MG
76659-874EA 87 USD
76659-874 76659-876 76659-878
5-(Aminomethyl)isoindoline-1,3-dione hydrochloride 97%
5-(Aminomethyl)isoindoline-1,3-dione hydrochloride

Precaución: for Research Use Only

Formula: C₉H₉ClN₂O₂
MDL Number: MFCD34165691
CAS Number: 2682112-48-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR