N-(4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)acrylamide 98%

Supplier: Ambeed
Warning

A1372127-1G A1372127-250MG A1372127-5G
76659-848EA 84 USD
76659-848 76659-850 76659-852
N-(4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)acrylamide 98%
N-(4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)acrylamide

Precaución: for Research Use Only

Formula: C₁₅H₂₀BNO₃
CAS Number: 434898-99-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR