(4S,4'S)-2,2'-(1,3-Bis(4-(tert-butyl)phenyl)propane-2,2-diyl)bis(4-(tert-butyl)-4,5-dihydrooxazole) 98%

Supplier: Ambeed
Warning

A1372076-100MG A1372076-1G A1372076-250MG
76659-842EA 94.43 USD
76659-842 76659-844 76659-846
(4S,4'S)-2,2'-(1,3-Bis(4-(tert-butyl)phenyl)propane-2,2-diyl)bis(4-(tert-butyl)-4,5-dihydrooxazole) 98%
(4S,4'S)-2,2'-(1,3-Bis(4-(tert-butyl)phenyl)propane-2,2-diyl)bis(4-(tert-butyl)-4,5-dihydrooxazole)

Precaución: for Research Use Only

Formula: C₃₇H₅₄N₂O₂
MW: 558.8369 g/mol
CAS Number: 2283330-44-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR