Coomassie brilliant blue G-250 BS
Supplier: Ambeed
|
Warning
|
Synonyms:
C.I. 42655
A1372057-100G
A1372057-5G
A1372057-25G
A1372057-500G
76659-838EA
91
USD
76659-838
76659-840
77621-914
77867-675
Coomassie brilliant blue G-250 BS
Coomassie brilliant blue G-250
Precaución: for Research Use Only
Formula:
C₄₇H₄₈N₃NaO₇S₂ MW: 854.04 g/mol |
MDL Number:
MFCD00078482 CAS Number: 6104-58-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...