(Ethoxycarbonylmethyl)triphenylphosphonium chloride 97%

Supplier: Ambeed
Warning

A1372051-100G A1372051-25G A1372051-500G
76659-828EA 57 USD
76659-828 76659-830 77621-912
(Ethoxycarbonylmethyl)triphenylphosphonium chloride 97%
(Ethoxycarbonylmethyl)triphenylphosphonium chloride

Precaución: for Research Use Only

Formula: C₂₂H₂₂ClO₂P
MDL Number: MFCD00011746
CAS Number: 17577-28-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR