2,3'-Dibromo-4'-fluoroacetophenone 97%

Supplier: Ambeed
Danger

A1372047-10G A1372047-1G
76659-824EA 114 USD
76659-824 76659-826
2,3'-Dibromo-4'-fluoroacetophenone 97%
2,3'-Dibromo-4'-fluoroacetophenone

Precaución: for Research Use Only

Formula: C₈H₅Br₂FO
MDL Number: MFCD07368759
CAS Number: 435273-49-7
UN: 3261
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR