(2-Chloro-4-(dimethylamino)phenyl)methanol 97%

Supplier: Ambeed
Warning

A1371935-100MG A1371935-250MG
76659-814EA 102.91 USD
76659-814 76659-816
(2-Chloro-4-(dimethylamino)phenyl)methanol 97%
(2-Chloro-4-(dimethylamino)phenyl)methanol

Precaución: for Research Use Only

Formula: C₉H₁₂ClNO
MW: 185.6507 g/mol
MDL Number: MFCD06202850
CAS Number: 85654-74-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR