Ni(tBustb)3 95%

Supplier: Ambeed
Danger

A1371820-1G A1371820-5G A1371820-250MG
76659-812EA 114.58 USD
76659-812 77549-664 77566-736
Ni(tBustb)3 95%
Ni(tBustb)3

Precaución: For research use only.

Formula: C₆₆H₈₄Ni
MW: 936.066 g/mol
MDL Number: MFCD32899499
CAS Number: 2468315-70-8
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR