L-Aspartic acid 98% 98atom%15N

Supplier: Ambeed
Warning

Synonyms: L-Aspartic acid-15N

A1371493-100MG A1371493-250MG
76659-792EA 161 USD
76659-792 76659-794
L-Aspartic acid 98% 98atom%15N
L-Aspartic acid

Precaución: for Research Use Only

Formula: C₄H₇NO₄
MDL Number: MFCD00055777
CAS Number: 3715-16-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR