4-Ethoxy-N-(5-nitrothiazol-2-yl)benzamide 97%

Supplier: Ambeed
Warning

A1371444-10MG A1371444-1MG A1371444-25MG A1371444-50MG A1371444-5MG
76659-782EA 33 USD
76659-782 76659-784 76659-786 76659-788 76659-790
4-Ethoxy-N-(5-nitrothiazol-2-yl)benzamide 97%
4-Ethoxy-N-(5-nitrothiazol-2-yl)benzamide

Precaución: for Research Use Only

Formula: C₁₂H₁₁N₃O₄S
MDL Number: MFCD01216148
CAS Number: 312608-54-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR