(S)-4-(tert-Butyl)-2-(pyridin-2-ylmethyl)-4,5-dihydrooxazole 97%

Supplier: Ambeed
Warning

A1371349-100MG A1371349-250MG
76659-766EA 52 USD
76659-766 77621-902
(S)-4-(tert-Butyl)-2-(pyridin-2-ylmethyl)-4,5-dihydrooxazole 97%
(S)-4-(tert-Butyl)-2-(pyridin-2-ylmethyl)-4,5-dihydrooxazole

Precaución: for Research Use Only

Formula: C₁₃H₁₈N₂O
MW: 218.2900 g/mol
MDL Number: MFCD34563690
CAS Number: 404844-77-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR