(S)-4-Benzyl-2-(pyrazin-2-yl)-4,5-dihydrooxazole 95%

Supplier: Ambeed
Warning

A1371326-100MG A1371326-1G A1371326-250MG
76659-718EA 106 USD
76659-718 76659-720 76659-722
(S)-4-Benzyl-2-(pyrazin-2-yl)-4,5-dihydrooxazole 95%
(S)-4-Benzyl-2-(pyrazin-2-yl)-4,5-dihydrooxazole

Precaución: for Research Use Only

Formula: C₁₄H₁₃N₃O
CAS Number: 1632140-86-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR