Methanesulfonato(diadamantyl-n-butylphosphino)-2'-amino-1,1'-biphenyl-2-yl)palladium(II) 97% (Oxide Impurities< 2%)

Supplier: Ambeed
Warning

A1371270-100MG A1371270-250MG A1371270-5G A1371270-1G
76659-706EA 18.04 USD
76659-706 76659-708 76659-710 77592-600
Methanesulfonato(diadamantyl-n-butylphosphino)-2'-amino-1,1'-biphenyl-2-yl)palladium(II) 97% (Oxide Impurities< 2%)
Methanesulfonato(diadamantyl-n-butylphosphino)-2'-amino-1,1'-biphenyl-2-yl)palladium(II)

Precaución: for Research Use Only

Formula: C₃₇H₅₂NO₃PPdS
MW: 728.27 g/mol
MDL Number: MFCD28144563
CAS Number: 1651823-59-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR