S-(+)-5,5'-Bis(diphenylphosphino)-2,2,2',2'-tetrafluoro-4,4'-bi-1,3-benzodioxole, dichloromethane adduct ≥98%, ee 99%

Supplier: Ambeed
Warning

Synonyms: (S)-DIFLUORPHOS™, (S)-5,5'-Bis(diphenylphosphino)-2,2,2',2'-tetrafluoro-4,4'-bi-1,3-benzodioxole

A1371114-100MG A1371114-1G A1371114-250MG A1371114-5G
76659-678EA 108.22 USD
76659-678 76659-680 76659-682 77597-008
S-(+)-5,5'-Bis(diphenylphosphino)-2,2,2',2'-tetrafluoro-4,4'-bi-1,3-benzodioxole, dichloromethane adduct ≥98%, ee 99%
S-(+)-5,5'-Bis(diphenylphosphino)-2,2,2',2'-tetrafluoro-4,4'-bi-1,3-benzodioxole, dichloromethane adduct

Precaución: for Research Use Only

MDL Number: MFCD05861607
CAS Number: 503538-70-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR