N,N-Bis[(1R)-(+)-phenylethyl]dibenzo[d,f][1,3,2]dioxaphosphepin-6-amine 98%

Supplier: Ambeed
Warning

A1371112-100MG A1371112-1G A1371112-250MG
76659-672EA 44.56 USD
76659-672 76659-674 76659-676
N,N-Bis[(1R)-(+)-phenylethyl]dibenzo[d,f][1,3,2]dioxaphosphepin-6-amine 98%
N,N-Bis[(1R)-(+)-phenylethyl]dibenzo[d,f][1,3,2]dioxaphosphepin-6-amine

Precaución: for Research Use Only

Formula: C₂₈H₂₆NO₂P
MW: 439.4853 g/mol
MDL Number: MFCD08277032
CAS Number: 500103-26-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR