4,6-Bis(diphenylphosphino)dibenzofuran 98%

Supplier: Ambeed
Warning

A1371107-1G A1371107-100MG
76659-670EA 379.81 USD
76659-670 77597-006
4,6-Bis(diphenylphosphino)dibenzofuran 98%
4,6-Bis(diphenylphosphino)dibenzofuran

Precaución: for Research Use Only

Formula: C₃₆H₂₆OP₂
MW: 536.5300 g/mol
MDL Number: MFCD00233867
CAS Number: 133850-81-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR