Methanesulfonato(2-dicyclohexylphosphino-2',6'-bis(dimethylamino)-1,1'-biphenyl)(2'-amino-1,1'-biphenyl-2-yl)palladium(II) 98%

Supplier: Ambeed
Warning

Synonyms: CPhos Pd G3

A1371102-250MG A1371102-5G A1371102-1G A1371102-100MG A1371102-25G
76659-666EA 15 USD
76659-666 77549-662 77566-730 77621-880 77867-645
Methanesulfonato(2-dicyclohexylphosphino-2',6'-bis(dimethylamino)-1,1'-biphenyl)(2'-amino-1,1'-biphenyl-2-yl)palladium(II) 98%
Methanesulfonato(2-dicyclohexylphosphino-2',6'-bis(dimethylamino)-1,1'-biphenyl)(2'-amino-1,1'-biphenyl-2-yl)palladium(II)

Precaución: For research use only.

Formula: C₄₁H₅₄N₃O₃PPdS
MDL Number: MFCD22417226
CAS Number: 1447963-73-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR