(R)-2-Benzyl-2,3-dihydrobenzo[d]imidazo[2,1-b]thiazole ≥97%, ee 99%

Supplier: Ambeed
Warning

A1371070-100MG A1371070-1G A1371070-250MG
76659-646EA 107.16 USD
76659-646 76659-648 76659-650
(R)-2-Benzyl-2,3-dihydrobenzo[d]imidazo[2,1-b]thiazole ≥97%, ee 99%
(R)-2-Benzyl-2,3-dihydrobenzo[d]imidazo[2,1-b]thiazole

Precaución: for Research Use Only

Formula: C₁₆H₁₄N₂S
MW: 266.3608 g/mol
CAS Number: 1415839-22-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR