(R)-2-(tert-Butyl)-2,3-dihydrobenzo[d]imidazo[2,1-b]thiazole 97%

Supplier: Ambeed
Warning

A1371066-100MG A1371066-1G A1371066-250MG
76659-640EA 100.79 USD
76659-640 76659-642 76659-644
(R)-2-(tert-Butyl)-2,3-dihydrobenzo[d]imidazo[2,1-b]thiazole 97%
(R)-2-(tert-Butyl)-2,3-dihydrobenzo[d]imidazo[2,1-b]thiazole

Precaución: for Research Use Only

Formula: C₁₃H₁₆N₂S
MW: 232.3445 g/mol
CAS Number: 1899061-68-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR