(S)-2-(tert-Butyl)-2,3-dihydrobenzo[d]imidazo[2,1-b]thiazole 97%

Supplier: Ambeed
Warning

A1371064-100MG A1371064-1G A1371064-250MG
76659-634EA 76.39 USD
76659-634 76659-636 76659-638
(S)-2-(tert-Butyl)-2,3-dihydrobenzo[d]imidazo[2,1-b]thiazole 97%
(S)-2-(tert-Butyl)-2,3-dihydrobenzo[d]imidazo[2,1-b]thiazole

Precaución: for Research Use Only

Formula: C₁₃H₁₆N₂S
MW: 232.3445 g/mol
CAS Number: 1213233-51-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR