(R)-2-Isopropyl-2,3-dihydrobenzo[d]imidazo[2,1-b]thiazole 97%

Supplier: Ambeed
Warning

A1371062-100MG A1371062-1G A1371062-250MG
76659-628EA 76.39 USD
76659-628 76659-630 76659-632
(R)-2-Isopropyl-2,3-dihydrobenzo[d]imidazo[2,1-b]thiazole 97%
(R)-2-Isopropyl-2,3-dihydrobenzo[d]imidazo[2,1-b]thiazole

Precaución: for Research Use Only

Formula: C₁₂H₁₄N₂S
MW: 218.3180 g/mol
CAS Number: 1415839-18-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR