Ethyl 3-methylpyrazolo[1,5-a]pyridine-2-carboxylate 97%

Supplier: Ambeed
Warning

A1370864-1G A1370864-250MG
76659-610EA 71 USD
76659-610 76659-612
Ethyl 3-methylpyrazolo[1,5-a]pyridine-2-carboxylate 97%
Ethyl 3-methylpyrazolo[1,5-a]pyridine-2-carboxylate

Precaución: for Research Use Only

Formula: C₁₁H₁₂N₂O₂
MW: 204.2252 g/mol
MDL Number: MFCD32066547
CAS Number: 1398584-48-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR