16-Methoxy-16-oxohexadecanoic acid 98%

Supplier: Ambeed
Warning

A1370671-1G A1370671-250MG A1370671-5G A1370671-25G
76659-606EA 25 USD
76659-606 76659-608 77549-658 77597-004
16-Methoxy-16-oxohexadecanoic acid 98%
16-Methoxy-16-oxohexadecanoic acid

Precaución: for Research Use Only

Formula: C₁₇H₃₂O₄
MDL Number: MFCD00673467
CAS Number: 18451-85-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR