Azo rubine 50% (dye content)

Supplier: Ambeed
Warning

A1370646-100G A1370646-25G A1370646-500G
76659-602EA 7 USD
76659-602 76659-604 77621-870
Azo rubine 50% (dye content)
Azo rubine

Precaución: for Research Use Only

Formula: C₂₀H₁₄N₂O₇S₂
MDL Number: MFCD00003978
CAS Number: 3567-69-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR