(S)-3-Amino-1-benzylpiperidine 96%

Supplier: Ambeed
Warning

Synonyms: (S)-1-Benzylpiperidin-3-amine

A1370638-1G A1370638-25G A1370638-5G
76659-596EA 23.34 USD
76659-596 76659-598 76659-600
(S)-3-Amino-1-benzylpiperidine 96%
(S)-3-Amino-1-benzylpiperidine

Precaución: for Research Use Only

Formula: C₁₂H₁₈N₂
MW: 190.29 g/mol
MDL Number: MFCD03093375
CAS Number: 168466-85-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR