Diethyl ((4-bromophenyl)difluoromethyl)phosphonate 97%

Supplier: Ambeed
Warning

A1370574-1G A1370574-250MG A1370574-5G
76659-590EA 40 USD
76659-590 76659-592 76659-594
Diethyl ((4-bromophenyl)difluoromethyl)phosphonate 97%
Diethyl ((4-bromophenyl)difluoromethyl)phosphonate

Precaución: for Research Use Only

Formula: C₁₁H₁₄BrF₂O₃P
CAS Number: 177284-56-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR