Monoethyl dodecanedioate 97%

Supplier: Ambeed
Warning

Synonyms: 11-Ethoxycarbonyldodecanoic acid, 12-Ethoxy-12-oxododecanoic acid, Dodecanedioic Acid Monoethyl Ester, Ethyl Hydrogen Dodecanedioate

A1370556-10G A1370556-1G
76659-586EA 225.98 USD
76659-586 76659-588
Monoethyl dodecanedioate 97%
Monoethyl dodecanedioate

Precaución: for Research Use Only

Formula: C₁₄H₂₆O₄
MW: 258.36 g/mol
MDL Number: MFCD00191643
CAS Number: 66003-63-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR