N2,N6-Dihydroxypyridine-2,6-bis(carboximidamide) 97%

Supplier: Ambeed
Warning

Synonyms: N2,N'6-dihydroxypyridine-2,6-bis(carboximidamide)

A1370554-1G A1370554-250MG
76659-582EA 203.7 USD
76659-582 76659-584
N2,N6-Dihydroxypyridine-2,6-bis(carboximidamide) 97%
N2,N6-Dihydroxypyridine-2,6-bis(carboximidamide)

Precaución: for Research Use Only

Formula: C₇H₉N₅O₂
MW: 195.1787 g/mol
MDL Number: MFCD01239956
CAS Number: 50545-09-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR