2-Anthracenecarboxylic acid 98%

Supplier: Ambeed
Warning

Synonyms: 2-Anthroic Acid, 2-Anthracenecarboxylic Acid

A1370544-100MG A1370544-250MG A1370544-1G
76659-578EA 22.28 USD
76659-578 76659-580 77621-868
2-Anthracenecarboxylic acid 98%
2-Anthracenecarboxylic acid

Precaución: for Research Use Only

Formula: C₁₅H₁₀O₂
MW: 222.24 g/mol
MDL Number: MFCD00142865
CAS Number: 613-08-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR