(4-(Hydrazinylmethyl)phenyl)(pyrrolidin-1-yl)methanone hydrochloride 97%

Supplier: Ambeed
Warning

A1370464-1G A1370464-250MG
76659-570EA 197.33 USD
76659-570 76659-572
(4-(Hydrazinylmethyl)phenyl)(pyrrolidin-1-yl)methanone hydrochloride 97%
(4-(Hydrazinylmethyl)phenyl)(pyrrolidin-1-yl)methanone hydrochloride

Precaución: for Research Use Only

Formula: C₁₂H₁₈ClN₃O
MW: 255.7438 g/mol
MDL Number: MFCD33395911
CAS Number: 2682112-51-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR