(4-(1,2,4,5-Tetrazin-3-yl)phenyl)methanamine hydrochloride 85%

Supplier: Ambeed
Warning

A1370300-1G A1370300-250MG A1370300-50MG A1370300-100MG A1370300-5G
76659-554EA 836 USD
76659-554 76659-556 76659-558 77621-864 77867-638
(4-(1,2,4,5-Tetrazin-3-yl)phenyl)methanamine hydrochloride 85%
(4-(1,2,4,5-Tetrazin-3-yl)phenyl)methanamine hydrochloride

Precaución: for Research Use Only

Formula: C₉H₁₀ClN₅
MDL Number: MFCD23097050
CAS Number: 1416711-59-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR