2-(4-Methylpiperidin-1-yl)propanoic acid hydrochloride 97%
Supplier: Ambeed
|
Warning
|
A1369656-250MG
A1369656-5G
76659-544EA
46.68
USD
76659-544
76659-546
2-(4-Methylpiperidin-1-yl)propanoic acid hydrochloride 97%
2-(4-Methylpiperidin-1-yl)propanoic acid hydrochloride
Precaución: for Research Use Only
Formula:
C₉H₁₈ClNO₂ MW: 207.6977 g/mol |
MDL Number:
MFCD33022462 CAS Number: 2682112-38-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...