Ethyl-2-oxobutanoate 96%

Supplier: Ambeed
Warning

Synonyms: Ethyl 2-oxobutanoate, Ethyl 2-oxobutyrate

A1369624-100G A1369624-1G A1369624-25G A1369624-500G A1369624-5G
76659-534EA 318 USD
76659-534 76659-536 76659-538 76659-540 76659-542
Ethyl-2-oxobutanoate 96%
Ethyl-2-oxobutanoate

Precaución: for Research Use Only

Formula: C₆H₁₀O₃
MW: 130.143 g/mol
MDL Number: MFCD03265336
CAS Number: 15933-07-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR