rel-(1R,2R)-2-(4-(Dimethylamino)phenyl)cyclopropanecarboxylic acid 97%

Supplier: Ambeed
Warning

A1369460-100MG A1369460-1G A1369460-250MG
76659-516EA 96.55 USD
76659-516 76659-518 76659-520
rel-(1R,2R)-2-(4-(Dimethylamino)phenyl)cyclopropanecarboxylic acid 97%
rel-(1R,2R)-2-(4-(Dimethylamino)phenyl)cyclopropanecarboxylic acid

Precaución: for Research Use Only

Formula: C₁₂H₁₅NO₂
MW: 205.2530 g/mol
CAS Number: 2307771-93-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR